Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel™ Phosphate Buffer w/MgCl 90mL
Thermo Scientific™ Remel Phosphate Buffer w/MgCl 90mL (pH 7.2 stock solution) formulation is used in water and wastewater testing as referenced in APHA and EPA test methods.
Supplier: Thermo Scientific™ R23702
This item is not returnable.
View return policy
Description
Thermo Scientific™ Remel Phosphate Buffer w/MgCl 90mL (pH 7.2 stock solution) formulation is used in water and wastewater testing as referenced in APHA and EPA test methods.
- Dilution Bottles are leakproof, wide mouth PET plastic with tear away sterility strip and easy, one-hand opening hinged lid
- Gamma irradiated
- Cap color-coded (gold) for easy and accurate visual identification
Specifications
Phosphate Buffer w/MgCl 90mL | |
Buffer Solution | |
72 x 90 mL |
Yellow Cap - 90mL/Bottle | |
72/Case |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction