Learn More
Thermo Scientific™ Remel™ Phosphate Buffer w/MgCl2 (pH 7.2) (Buffered Water) APHA Formulation
The Thermo Scientific™ Remel Phosphate Buffer with MgCl2 (pH 7.2) APHA Formulation is used as a diluent in microbial limit testing of food and water.
$223.00 - $1191.00
Catalog Number | Mfr. No. | dimFormat | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | dimFormat | Quantity | Price | Quantity & Availability | ||||
R112531
|
Thermo Scientific™
R112531 |
10 x 20 mL |
Pack of 10 for $276.00
|
|
|||||
R118610
|
Thermo Scientific™
R118610 |
100 x 9 mL |
Case of 100 for $1,191.00
|
|
|||||
R112530
|
Thermo Scientific™
R112530 |
10 x 20 mL |
Pack of 10 for $223.00
|
|
|||||
R112522
|
Thermo Scientific™
R112522 |
10 x 90 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||||
Description
Thermo Scientific™ Remel Phosphate Buffer with MgCl2 (pH 7.2) (Buffered Water) APHA Formulation is used as a diluent in microbial limit testing of food and water.
Ordering Information:
REF R112530 has open crimp seal. REF R112531 has complete crimp seal.
Specifications
Buffer Solution |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.