Learn More
Thermo Scientific™ Remel™ Phosphate Buffered Saline (Cold Enrichment Broth) pH 7.6 Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel Phosphate Buffered Saline (Cold Enrichment Broth) pH 7.6 is for the cultivation of Yersinia enterocolitica.
$217.88 - $1039.98
Specifications
Description Phosphate Buffered Saline
Format 15mm x 103mm Tube
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
R062582 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R062582
20 x 5mL Pack of 20 for $217.88
Add to cart
R062580 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R062580
100 x 5mL Case of 100 for $1,039.98
Add to cart
Description
Description
Thermo Scientific™ Remel Phosphate Buffered Saline (Cold Enrichment Broth) pH 7.6 is for the cultivation of Yersinia enterocolitica.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Phosphate Buffered Saline
15mm x 103mm Tube