Learn More
Thermo Scientific™ Remel™ PRAS Chopped Meat Broth (VPI) (Pre-Reduced Anaerobic Sterilized) (Chopped Meat Medium) Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel PRAS Chopped Meat Broth (VPI) (Pre-Reduced Anaerobic Sterilized) (Chopped Meat Medium) is used for cultivation of anaerobic bacteria.
$329.70
Specifications
Description PRAS Chopped Meat Broth
Packaging 20/Pk.
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
R05035 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R05035
20 x 7.5mL Pack of 20 for $329.70
Add to cart
Description
Description
Thermo Scientific™ Remel PRAS Chopped Meat Broth (VPI) (Pre-Reduced Anaerobic Sterilized) (Chopped Meat Medium) is used for cultivation of anaerobic bacteria.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
PRAS Chopped Meat Broth
20/Pk.