Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel™ Rappaport-Vassiliadis Salmonella Enrichment Broth MLT
Thermo Scientific™ Remel Rappaport-Vassiliadis Salmonella Enrichment Broth MLT facilitates selective enrichment of Salmonella spp.
$393.50 - $1689.00
Specifications
Certifications/Compliance | Industrial Reference: USP. |
---|---|
Description | Rappaport-Vassiliadis Salmonella Enrichment Broth MLT |
Format | 16mm x 125mm Tube |
Product Type | Microbiology Media |
Description
Thermo Scientific™ Remel Rappaport-Vassiliadis Salmonella Enrichment Broth MLT facilitates selective enrichment of Salmonella spp.
Specifications
Industrial Reference: USP. | |
Rappaport-Vassiliadis Salmonella Enrichment Broth MLT | |
16mm x 125mm Tube | |
Microbiology Media |
Not intended for IVD use.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title