Learn More
Thermo Scientific™ Remel™ SABHI™ Agar w/Chloramphenicol, Cycloheximide Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel SABHI Agar w/Chloramphenicol, Cycloheximide is for the cultivation, isolation of pathogenic fungi.
$129.15 - $1125.60
Specifications
Description SABHI Agar w/Chloramphenicol, Cycloheximide (Sabouraud Dextrose w/Brain Heart Infusion Agar)
View More Specs
Products 2
Catalog Number Mfr. No. Format Price Quantity & Availability  
Catalog Number Mfr. No. Format Price Quantity & Availability  
R01754 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R01754
Monoplate Deep Pour
Pack of 10 for $129.15
Add to cart
R08697
View Documents
Thermo Scientific™
R08697
20mm x 113mm Tube
Case of 100 for $1,125.60
Add to cart
Description
Description
Thermo Scientific™ Remel SABHI Agar w/Chloramphenicol, Cycloheximide is for the cultivation, isolation of pathogenic fungi.
Chloramphenicol inhibits some gram-positive and gram-negative bacteria. Cycloheximide inhibits rapidly growing saprophytic fungi.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
SABHI Agar w/Chloramphenicol, Cycloheximide (Sabouraud Dextrose w/Brain Heart Infusion Agar)