Learn More
Thermo Scientific™ Sabouraud Dextrose Agar, Emmons w/Chloramphenicol, Gentamicin Available on GSA/VA Contract for Federal Government customers only.
Isolate and cultivate pathogenic yeast and molds from clinical specimens with this selective, ready-to-use medium.
$128.84 - $310.28
Specifications
Description Sabouraud Dextrose Agar, Emmons w/Chloramphenicol, Gentamicin
View More Specs
Products 2
Catalog Number Mfr. No. Format Price Quantity & Availability  
Catalog Number Mfr. No. Format Price Quantity & Availability  
R01772 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R01772
Monoplate Deep Pour Pack of 10 for $128.84
Add to cart
R08742
Accessories View Documents
Thermo Scientific™
R08742
20mm x 113mm Tube Pack of 20 for $310.28
Add to cart
Description
Description
Isolate and cultivate pathogenic yeast and molds from clinical specimens with Thermo Scientific™ Remel™ Sabouraud Dextrose Agar (Emmons) prepared media plates. Sabouraud Dextrose Agar (Emmons) is a modification of the original medium developed by Sabouraud in 18921. Emmons neutralized the pH, increasing it to 7.0, and reduced the concertation of dextrose from 40 to 20 g/L2. The addition of antibiotics in the medium makes the medium inhibitory to bacterial overgrowth while allowing isolation of pathogenic fungi.
Use Sabouraud Dextrose Agar (Emmons) for primary isolation and cultivation of pathogenic yeasts and molds from clinical specimens.
  • Selective: selective agents chloramphenicol and gentamicin are inhibitory to bacterial overgrowth while allowing isolation of pathogenic fungi.
  • Suitable for isolation of pathogenic fungi from a material containing a large amount of other fungi or bacteria.
  • Ready prepared convenience of prepared media.
Casein and meat peptones supply nitrogenous compounds and amino acids necessary for the growth of fungi. Dextrose is an energy source which serves to support the growth of fungi. Chloramphenicol and gentamicin are broad-spectrum antibiotics which inhibit a range of Gram-positive and Gram-negative bacteria.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Sabouraud Dextrose Agar, Emmons w/Chloramphenicol, Gentamicin