Learn More
Thermo Scientific™ Remel™ Sabouraud Dextrose Agar, Emmons w/Gentamicin
Thermo Scientific™ Remel Sabouraud Dextrose Agar, Emmons with Gentamicin is used for the selective isolation of yeast and fungi.
$134.00
Specifications
Description Sabouraud Dextrose Agar, Emmons w/Gentamicin
View More Specs
Products 1
Catalog Number Mfr. No. Format Price Quantity & Availability  
Catalog Number Mfr. No. Format Price Quantity & Availability  
R01776
Accessories View Documents
Thermo Scientific™
R01776
Monoplate Deep Pour
Pack of 10 for $134.00
Add to cart
Description
Description
Thermo Scientific™ Remel Sabouraud Dextrose Agar, Emmons with Gentamicin is used for the selective isolation of yeast and fungi.
  • Gentamicin inhibits some gram-negative bacteria
Specifications
Specifications
Sabouraud Dextrose Agar, Emmons w/Gentamicin