Learn More
Thermo Scientific™ Schaedler Broth
Cultivate fastidious aerobic and anaerobic bacteria using Schaedler Broth.
Supplier:  Thermo Scientific™ R064482
 View more versions of this product
Catalog No. R064482
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
Cultivate fastidious aerobic and anaerobic bacteria using Thermo Scientific™ Remel™ Schaedler Broth. In 1965, Schaedler, Dubos, and Costello formulated a medium for isolation of aerobic and anaerobic microorganisms from the gastrointestinal tract of mice1. In 1974, Stalons, Thornsberry, and Dowell compared nine broth media in varied CO2 atmospheres for the ability to support the growth of anaerobic bacteria2. Schaedler Broth was found to be most effective of the medium tested and demonstrated superior growth of anaerobic microorganisms.
In Schaedler Broth agar is removed and has the same formulation as Schaedler Agar. Mata et al. modified the Schaedler formulation by adjusting the concentrations of sodium chloride and peptone, reducing the amount of glucose to avoid interference with
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Schaedler Broth
15mm x 103mm Tube
Microbiology Media
20 x 5 mL
SDS
Documents
Documents
Product Certifications
Accessories