Learn More
Thermo Scientific™ Remel™ SIM Medium (Sulfide, Indole, Motility) Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel SIM Medium (Sulfide, Indole, Motility) differentiates enteric bacilli on the basis of sulfide production, indole formation, and motility.
$183.02 - $903.00
Specifications
Certifications/Compliance Industrial Reference: BAM.
Description SIM Medium (Sulfide; Indole; Motility)
Format 15mm x 103mm Tube
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
R064542 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R064542
20 x 5mL Pack of 20 for $183.02
Add to cart
R064540 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R064540
100 x 5mL Case of 100 for $903.00
Add to cart
Description
Description
Thermo Scientific™ Remel SIM Medium (Sulfide, Indole, Motility) is for the differentiation of enteric bacilli on the basis of sulfide production, indole formation, and motility.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Industrial Reference: BAM.
SIM Medium (Sulfide; Indole; Motility)
15mm x 103mm Tube