Learn More
Thermo Scientific™ SXT Blood Agar
Selectively isolate beta-hemolytic Group A Streptococci from throat cultures and upper respiratory specimens using SXT Blood Agar.
Supplier: Thermo Scientific™ R01861
Description
Selectively isolate beta-hemolytic group A streptococci from throat cultures and upper respiratory specimens using Thermo Scientific™ Remel™ SXT Blood Agar. Pacifico at al. demonstrated the isolation of β-hemolyic streptococci is improved with the use of a selective medium which inhibits oropharyngeal commensal microbial flora1. In 1977, Gunn et al. introduced a selective medium containing sulfamethoxazole and trimethoprim in a tryptic soy agar base supplemented with 5% sheep blood2. In 1979, Kurzynski et al. proved that streptococci were more frequently isolated on SXT sheep blood agar3.
Sulfamethoxazole and trimethoprim suppress growth of streptococcal groups C, F, and G and Streptococci viridans, most Enterobacteriaceae, Neisseria, and some Pseudomonas spp.Specifications
SXT Blood Agar w/ sulfamethoxazole, trimethoprim | |
10mm x 100mm Spacesaver Plate | |
Agar | |
15/Pk. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.