Learn More
Thermo Scientific™ Remel™ TAT Broth
Thermo Scientific™ Remel TAT Broth (w/Polysorbate 20) is used for sterility testing of highly viscous substances, such as salves, ointments and cosmetics.
$207.00 - $226.28
Specifications
Description TAT Broth, w/Polysorbate 20
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
R112611
Accessories View Documents
Thermo Scientific™
R112611
10 x 90mL Case of 10 for $226.28
Add to cart
R112612
Accessories View Documents
Thermo Scientific™
R112612
1000mL Each for $207.00
Add to cart
Description
Description
Thermo Scientific™ Remel TAT Broth (w/Polysorbate 20) is used for sterility testing of highly viscous substances, such as salves, ointments and cosmetics.
Specifications
Specifications
TAT Broth, w/Polysorbate 20
Not intended for IVD use.