Learn More
Thermo Scientific™ Remel™ TB Decolorizer, Truant-Moore Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Remel TB Decolorizer, Truant-Moore is for TB Auramine O and TB Auramine-Rhodamine T fluorescent stains.
$46.25 - $122.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
R40207 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R40207
5/Pk.
Pack of 5 for $122.00
Add to cart
R40107 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R40107
Each
Each for $46.25
Add to cart
Description
Description
Thermo Scientific™ Remel TB Decolorizer, Truant-Moore is for TB Auramine O and TB Auramine-Rhodamine T fluorescent stains.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.