Learn More
Thermo Scientific™ Remel™ Tetrathionate Broth Base Available on GSA/VA Contract for Federal Government customers only.
Remel Tetrathionate Broth Base is selective enrichment medium for use with iodine for the recovery of Salmonella species.
$120.23 - $523.43
Specifications
Description Tetrathionate Broth Base
Format Powder
Packaging Ea.
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
R454822 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R454822
500g Each for $120.23
Add to cart
R454824 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R454824
2.5kg Each for $523.43
Add to cart
Description
Description
Remel Tetrathionate Broth Base is selective enrichment medium for use with iodine for the recovery of Salmonella species.
Also available: Brilliant Green for Tetrathionate, Part No. R114080 and Iodine For Tetrathionate, Part No. R114350.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Tetrathionate Broth Base
Powder
Ea.
For Laboratory Use Only