Learn More
Thermo Scientific™ Remel™ Tetrathionate Broth
Thermo Scientific™ Remel Tetrathionate Broth is used for selective enrichment and isolation of Salmonella species.
Manufacturer:  Thermo Scientific™ R112630
 View more versions of this product
Catalog No. R112630
Add to cart
This item is not returnable. View return policy
Description
Description
Thermo Scientific™ Remel Tetrathionate Broth is used for selective enrichment and isolation of Salmonella species.
Specifications
Specifications
Tetrathionate Broth
10 x 90mL
Industrial Reference: AOAC, BAM, COMPF, SMWW, USDA, USP.
Broth
WMB Lug - 90mL
10/Pk.
SDS
Documents
Accessories
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Requires addition of Iodine Solution, REF R21228, REF R114350, or REF R114351. REF R112634 and REF R112630 not intended for IVD use.