Learn More
Thermo Scientific™ Remel™ Thioglycollate Medium with Dextrose Available on GSA/VA Contract for Federal Government customers only.
General-purpose growth medium for cultivation of aerobic and anaerobic microorganisms.
$136.82 - $1518.30
Specifications
Type Media
View More Specs
Products 8
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Format Quantity Price Quantity & Availability  
R064700 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R064700
15mm x 103mm Tube 100 x 9mL Case of 100 for $1,162.77
Add to cart
R064702 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R064702
15mm x 103mm Tube 20 x 9mL Pack of 20 for $136.82
Add to cart
R07176
Accessories View Documents
Thermo Scientific™
R07176
16mm x 125mm Tube 100 x 10mL Case of 100 for $770.70
Add to cart
R07178 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R07178
16mm x 125mm Tube 20 x 10mL Pack of 20 for $168.11
Add to cart
R08850 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R08850
20mm x 113mm Tube 100 x 10mL Case of 100 for $1,518.30
Add to cart
R08852
Accessories View Documents
Thermo Scientific™
R08852
20mm x 113mm Tube 20 x 10mL Pack of 20 for $154.46
Add to cart
R08856
Accessories View Documents
Thermo Scientific™
R08856
20mm x 113mm Tube 100 x 18mL Case of 100 for $821.63
Add to cart
R08858
View Documents
Thermo Scientific™
R08858
F - 18mL 20 x 18mL Pack of 20 for $142.70
Add to cart
Description
Description
Thermo Scientific™ Remel Thioglycollate Medium with Dextrose (without Indicator) is a general-purpose growth medium for cultivation of aerobic and anaerobic microorganisms.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Media