Learn More
Thermo Scientific™ Thioglycollate Medium w/Dextrose
Cultivate aerobes and anaerobes using general purpose Thioglycollate Medium w/Dextrose.
Supplier: Thermo Scientific™ R064702
Description
Cultivate aerobes and anaerobes using Thermo Scientific™ Remel™ Thioglycollate Medium w/Dextrose. This general-purpose medium was first described by Brewer in 19401 and consists of a small amount of agar and sodium thioglycolate, a reducing substance, to initiate the growth of anaerobes.
Thioglycollate Medium w/Dextrose is a liquid medium recommended for use as a general purpose media. Sodium thioglycollate is the reducing agent which removes molecular oxygen from the medium and prevents the accumulation of peroxides which may be letha
Specifications
Thioglycollate Medium; 9mL; 15 x 103mm tube | |
15mm x 103mm Tube | |
Microbiology Media | |
20 x 9 mL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.