Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel™ Tryptic Soy Broth
The Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.
Supplier: Thermo Scientific™ R112730
This item is not returnable.
View return policy
Description
Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.
- Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia) and for use as general purpose enrichment broth
Specifications
Industrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP. | |
WMB Lug - 100mL | |
10/Pk. |
Tryptic Soy Broth | |
Microbiology Media | |
10 x 100 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
REF numbers beginning with R11 are not intended for IVD use.
Spot an opportunity for improvement?
Provide Content Correction