Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Urea Agar Available on GSA/VA Contract for Federal Government customers only.

Thermo Scientific™ Remel Urea Agar (Christensens) facilitates detection of urea hydrolysis by microorganisms.

$185.96 - $1025.33

Specifications

Certifications/Compliance Industrial Reference: BAM.
Description Urea Agar
Format 15mm x 103mm Tube - Slant
View More Specs

Products 2
Catalog Number Mfr. No. Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Price Quantity & Availability  
R065210 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R065210
100/Pk. Case of 100 for $1,025.33
Add to cart
 
R065212 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
R065212
20/Pk. Pack of 20 for $185.96
Add to cart
 
Description

Description

Thermo Scientific™ Remel Urea Agar (Christensens) facilitates detection of urea hydrolysis by microorganisms.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Industrial Reference: BAM.
Urea Agar
15mm x 103mm Tube - Slant
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit