Learn More
Thermo Scientific™ Urea Broth
Differentiate Enterobacteriaceae based on urea hydrolysis using ready-to-use liquid medium Urea Broth (Stuart’s).
$442.00
Specifications
Description | Urea Broth |
---|---|
Format | 15mm x 103mm Tube |
Product Type | Microbiology Media |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
R065232
|
Thermo Scientific™
R065232 |
20 x 2 mL |
Pack of 20 for $442.00
|
|
|||||
R065230
|
Thermo Scientific™
R065230 |
100 x 2 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||||
Description
Differentiate genera within the Enterobacteriaceae on the basis of urease production with Thermo Scientific™ Remel™ Urea Broth (Stuart’s). Urea Broth (Stuart’s) was developed by Stuart et al. to differentiate Proteus from other genera within the Enterobacteriaceae1,2. This medium is designed such that it detects urease activity in organisms which produce ammonia in a highly buffered medium with limited nutrients. Such organisms include Proteus spp., Morganella morganii, Providencia rettgeri, and urease-positive Providencia stuartii3.
Urea Broth (Stuart’s) is a liquid medium used to detect microorganisms on the basis of urease production. It is a medium with high buffering capacity and limited nutrients. Other genera within the Enterobacteriaceae which hydrolyze urea, will not do so in this medium.
- Ready to use – Liquid medium.
- Easy to interpret – An intense pink-red color throughout the broth indicates a positive test. No color change, the broth remains yellow-orange in the case of a negative test.
This medium contains yeast extract that provides essential growth factors required by strong urease-positive organims such as Proteus spp., M. morganii, and some species of Providencia. Urea is the substrate that can be hydrolyzed. Disodium and monopotassium phosphates provide the buffering system. Phenol red acts as the pH indicator.
Organisms which possess the urease enzyme hydrolyze urea and produce an alkaline by-product (ammonia) which is detected when the phenol red indicator changes color from yellow-orange to pink.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Urea Broth | |
15mm x 103mm Tube | |
Microbiology Media |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.