Learn More
Thermo Scientific™ Remel™ Violet Red Bile Agar
The Thermo Scientific™ Remel Violet Red Bile Agar is used for selective isolation and detection of coliform microorganisms in water, foods and dairy products.
Manufacturer:  Thermo Scientific™ R112872
Catalog No. R112872
Add to cart
This item is not returnable. View return policy
Description
Description
Thermo Scientific™ Remel Violet Red Bile Agar is used for selective isolation and detection of coliform microorganisms in water, foods, milk and other dairy products.
Specifications
Specifications
Violet Red Bile Agar
Agar
BB - 500mL
10/Pk.
Agar
10 x 500mL
Industrial Reference: BAM, COMPF, IDF.
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Not intended for IVD use.