Learn More
Thermo Scientific™ Wellcogen™ Streptococcus pneumoniae Rapid Latex Agglutination Test, Wellcogen S. pneumoniae Kit
Rapidly detect capsular antigens to Streptococcus pneumoniae present in human body fluids or blood cultures by latex agglutination.
Supplier: Thermo Scientific™ R30859001
Description
Wellcogen™ S. pneumoniae is a rapid latex test for use in the qualitative detection of capsular antigen from Streptococcus pneumoniae (Pneumococcus), present in body fluids as a consequence of infection or in blood cultures. NOTE: Tests performed directly on clinical specimens are intended for screening purposes and should augment, not replace, culture procedures. Results must be used in conjunction with other data; e.g. symptoms, results of other tests, clinical impressions etc.
Use this rapid latex test in the qualitative detection of capsular antigen from Streptococcus pneumoniae (Pneumococcus) present in body fluids, as a consequence of infection, or in blood cultures.The reagent consists of polystyrene latex particles which have been coated with antibodies purified from an omnivalent serum which reacts with all the recognised serological types of pneumococci. These latex particles agglutinate in the presence of sufficient homologous antigen.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
2°C to 8°C | |
Yes | |
Streptococcus Test Kit |
Wellcogen Streptococcus pneumoniae Test Kit | |
S. pneumoniae | |
30 Tests/Kit |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.