Learn More
Thermo Scientific™ Cryptosporidium Controls
Validate the test procedures of Thermo Scientific™ Xpect™ Remel™ Giardia/Cryptosporidium control test kits with Thermo Scientific Xpect Remel Cryptosporidium Controls.
Supplier: Thermo Scientific™ R2451030
Description
Validate the test procedures of Thermo Scientific™ Xpect™ Remel™ Giardia/Cryptosporidium control test kits with Thermo Scientific Xpect Remel Cryptosporidium Controls These kits contain positive and negative controls and are intended for QC purposes, to confirm that the test has been performed satisfactorily and can produce the expected result.
Cryptosporidium is a protozoan pathogen of the Apicomplexa phylum and causes a diarrheal illness called cryptosporidiosis. Cryptosporidiosis is typically an acute short-term infection but can become severe and non-resolving in children and immunocompromised individuals such as AIDS patients.
This kit contains Cryptosporidium positive control which is derived from 10% Formalin or SAF fixative stool samples of human or animal origin which are positive for Cryptosporidium; Giardia positive control derived from 10% formalin or SAF fixed stool samples which are positive for Giardia and Giardia/Cryptosporidium negative control which is derived from 10% Formalin or SAF fixative stool samples, which are negative for Giardia and Cryptosporidium.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Xpect™ Cryptosporidium Control Kit | |
1 positive vial and 1 negative vial | |
2 Vials/kit |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.