Learn More
Thermo Scientific™ Remel™ Yeast Nitrogen Base w/o Amino Acids
Thermo Scientific™ Remel Yeast Nitrogen Base w/o Amino Acids is used for the identification of yeast based on amino acid and carbohydrate requirements.
Supplier: Thermo Scientific™ R459942
Description
Thermo Scientific™ Remel Yeast Nitrogen Base w/o Amino Acids is used for the identification of yeast based on amino acid and carbohydrate requirements.
Specifications
Yeast Nitrogen Base w/o Amino Acids | |
Powder | |
Dehydrated Culture Media | |
500 g |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Laboratory Use Only