Learn More
Thermo Scientific™ Remote Alarms for Ultra-Low Temperature Freezers
Increase sample security by using these remote alarms for ultra-low temperature freezers—designed for areas where an alarm may not be readily heard.
$1164.00
Specifications
Description Remote Alarm with Audible and Visual Signal
View More Specs
Products 1
Catalog Number Mfr. No. Voltage Price Quantity & Availability  
Catalog Number Mfr. No. Voltage Price Quantity & Availability  
13-986-32
View Documents
Thermo Scientific™
56121A
120 V Each for $1,164.00
Add to cart
Description
Description
Increase sample security by using Thermo Scientific™ Remote Alarms for Ultra-Low Temperature Freezers — designed for areas where an alarm may not be readily heard.
Features:
  • Lightweight units attach to a wall or cabinet
  • Operate on line voltage
  • Switch to a battery backup if a power failure occurs
  • Can be connected easily to a temperature/power monitor and remote alarm contacts
  • Complete with audible alarm silence switch and alarm ringback features
  • Alarm can be temporarily defeated, but system rings back as a reminder to reset when normal conditions are restored
  • Audible and visual signal to indicate alarm condition on monitored equipment
Specifications
Specifications
Remote Alarm with Audible and Visual Signal