Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals Renal Load Diet, Low, Powder

Low renal load diet. Contains purified high nitrogen casein.

Manufacturer:  MP Biomedicals 0296033050

 View more versions of this product

Catalog No. MP296033050


Add to cart

Description

Description

  • Low renal load diet
  • Contains purified high nitrogen casein, AIN-76 mineral mix, 1.2X vitamin, from normal concentration
Specifications

Specifications

1 ft.
0.3048 m
Animal Diet
Room Temperature
Yes
50 kg
Autoclavable
Non-sterile
Powder
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.