Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Renalase Mouse anti-Human, Clone: RNLS/1940, Novus Biologicals™

Mouse Monoclonal Antibody

$138.37 - $352.89

Specifications

Antigen Renalase
Clone RNLS/1940
Concentration 0.2 mg/ml
Dilution ELISA
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122806
View Documents
Novus Biologicals
NBP30766620UG
20 μg
Each for $138.37
Add to cart
 
NB122805
View Documents
Novus Biologicals
NBP30766610UG
100 μg
Each for $352.89
Add to cart
 
Description

Description

Renalase Monoclonal antibody specifically detects Renalase in Human samples. It is validated for ELISA
Specifications

Specifications

Renalase
0.2 mg/ml
ELISA
Purified
RUO
Human
C10orf59chromosome 10 open reading frame 59, EC 1.4, FLJ11218, MAO-C, Monoamine oxidase-C, RENALASE, renalase, FAD-dependent amine oxidase
Recombinant human Renalase protein fragment (around aa 34-235) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
RNLS/1940
ELISA
Unconjugated
Mouse
Lipid and Metabolism
10 mM PBS with 0.05% BSA
55328
IgG2b Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit