Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RENALASE Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 150031AP

Catalog No. 50-172-9901


Add to cart

Description

Description

This antibody is specific to RNLS.

Renalase is a flavin adenine dinucleotide-dependent amine oxidase that is secreted into the blood from the kidney.
Specifications

Specifications

RENALASE
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A7RDN6, Q5U2W9, Q5VYX0
C10orf59, MAO C, Monoamine oxidase C, RENALASE, RNLS
Rabbit
IgG
150 μL
-20°C
Primary
361751, 55328, 67795
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RNLS
Liquid
Rnls
RENALASE Fusion Protein Ag13061
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit