Learn More
RenalCellCarcinoma(gp200) Mouse anti-Human, Janelia Fluor 549, Clone: HD30, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP141003JF549
Catalog No. NBP14003549
Add to cart
Description
Description
RenalCellCarcinoma(gp200) Monoclonal antibody specifically detects RenalCellCarcinoma(gp200) in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
RenalCellCarcinoma(gp200)
HD30
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
LY75
Purified extracellular domain of human CD205-IgG1 Fc fusion protein.
Protein G purified
Dendritic Cell Markers, Ovarian Carcinoma Cell Markers, Renal Cell Carcinoma Cell Markers, Vision
Primary
4065
Human, Primate
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
CD205LY-75, CLEC13BLy-75, C-type lectin domain family 13 member B, DEC-205CD205 antigen, GP200-MR6, lymphocyte antigen 75
Mouse
IgG1
0.1mL
Store at 4°C in the dark.
Monoclonal
This antibody specifically recognises CD205, otherwise known as DEC205, a type I transmembrane glycoprotein and member of the macrophage mannose receptor family of C-type lectins, predominantly expressed by human dendritic cells.