Learn More
Drg International IncSupplier Diversity Partner Renin (Active) Elisa Ruo DFS Item
Manufacturer:  Drg International IncSupplier Diversity Partner EIA5125R
An enzyme immunoassay for the quantitative measurement of active renin in serum and EDTA plasma. This product is for Research Use Only. The DRG Renin ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) based on the sandwich principle. The microtiter wells are coated with a monoclonal [mouse] antibody directed towards a unique antigenic site of the human active Renin molecule. 
Catalog No. 50-194-7626