Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RENT1, Polyclonal, Bethyl Laboratories Non-distribution product as customer accommodation.

Goat Polyclonal Antibody

Manufacturer:  Bethyl Laboratories, IncSupplier Diversity Partner A300036AM

 View more versions of this product

Catalog No. 50-155-6774


Add to cart

Description

Description

The recommended shelf life for this product is 1 year from date of receipt.

This gene encodes a protein that is part of a post-splicing multiprotein complex involved in both mRNA nuclear export and mRNA surveillance. mRNA surveillance detects exported mRNAs with truncated open reading frames and initiates nonsense-mediated mRNA decay (NMD). When translation ends upstream from the last exon-exon junction, this triggers NMD to degrade mRNAs containing premature stop codons. This protein is located only in the cytoplasm. When translation ends, it interacts with the protein that is a functional homolog of yeast Upf2p to trigger mRNA decapping. Use of multiple polyadenylation sites has been noted for this gene.
Specifications

Specifications

RENT1
Polyclonal
TBS with 0.1% BSA and 0.09% sodium azide
Q92900, Q9EPU0
Regulator of nonsense transcripts 1; ATP-dependent helicase RENT1; Nonsense mRNA reducing factor 1; NORF1; Up-frameshift suppressor 1 homolog; hUpf1; RENT1; KIAA0221; UPF1;
Goat
IgG
100 μl
4° C
Primary
19704, 5976
Western Blot
Unconjugated
UPF1
liquid
Upf1
Between 250 and 300
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit