Learn More
Thermo Scientific™ Repair Kits for Thermo Scientific™ Plunger-in-Needle Syringes
Extend the life and performance, and reduce the cost per use of your syringes with the Thermo Scientific™ Repair Kit for Thermo Scientific Plunger-in-Needle Syringes.
$59.90
Specifications
Needle Gauge 23 G
View More Specs
Products 3
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
36-5RK-341
View Documents
Thermo Scientific™
365RK341
For PIN Syringe 365B0531 Each for $59.90
Add to cart
36-5RK-321
View Documents
Thermo Scientific™
365RK321
For PIN syringe 365A0331 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
36-5RK-311
View Documents
Thermo Scientific™
365RK311
For PIN syringe 365B0511 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Thermo Scientific™ Repair Kits for Thermo Scientific Plunger-in-Needle Syringes extend the life and performance and reduce cost per use of syringes.
  • Repair kits for plunger in needle syringes contain both the plunger and needle, which must be changed together
Includes: Plunger, 1; Needle, 1
Specifications
Specifications
23 G