Learn More
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Non-sterile
Corning™ Step-R™ Repeating Pipettor Syringe Tips, Non-sterile provide precise, positive-displacement dispensing of viscous or volatile liquids.
Manufacturer:  Corning™ 6626
 View more versions of this product
Catalog No. 07-201-481
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Polypropylene tips for use with the Corning™ Step-R™ Repeating Pipettor
  • Available in eight sizes from 0.05mL to 50mL
  • Tips use the direct displacement method, dedicated for use with viscous, infectious and volatile liquids
  • Reliable and repeatable results through automated, individual tests of every single tip
  • Certification of every batch is available on request
Specifications
Specifications
Yes
Polypropylene
25mL
100/PK
Non-sterile
Dispensing of viscous and volatile liquids
ISO8655