Learn More
Argos Technologies P-Vac Portable Aspirating System
Ideal for laboratory bench or laminar hood. Argos Technologies P-Vac Portable Vacuum System designed as a compact, flexible and portable liquid waste disposal system.
Manufacturer:  Argos Technologies PV010
 View more versions of this product
Catalog No. 03-391-218
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Designed to accommodate bottle top aspirating filter units, plastic vacuum bottles, glass Erlenmeyer flasks, or any device that accepts vacuum
  • Rechargeable lithium ion battery allows for use where inline vacuum source is not available
Specifications
Specifications
2900 mAh / 10.5 Wh
P-Vac aspiration pump
Yes
Lithium rechargeable battery
P-Vac replacement battery
1
3.6 V