Learn More
Fisherbrand™ Replacement Deuterium Lamp for Dionex UV and Visible HPLC Detector
Fisherbrand™ Replacement Deuterium Lamp for Dionex UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
Description
Fisherbrand™ Replacement Deuterium Lamp for Dionex UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
- Long life
- Drop-in OEM replacement
- Traceable by serial number
Order Info
Cat. No. 14-500-723 is comparable to Dionex part number 6080.2001, 054157.
Compliance
WEEE and ROHS compliant
Specifications
Replacement Detector Lamp | |
Deuterium | |
Dionex PDA-100, PDA-3000, AD-25 UV and Visible HPLC Detector |