Learn More
Fisherbrand™ Replacement Deuterium Lamp For Shimadzu UV and Visible HPLC Detector
Fisherbrand™ Replacement Deuterium Lamp For Shimadzu UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
Description
- Long life
- Drop-in OEM replacement
- Traceable by serial number
Order Info
Cat. No. 14-500-725 is comparable to Shimadzu part 228-34016-02.
Compliance
WEEE and ROHS compliant
Specifications
Replacement Detector Lamp | |
Deuterium | |
Shimadzu SPD-10A, AVP, AV, AVVP, M1 UV and Visible HPLC Detector |