Learn More
Fisherbrand™ Replacement Deuterium Lamp for Varian UV and Visible HPLC Detector
Fisherbrand™ Replacement Deuterium Lamp for Varian UV and Visible HPLC Detector has 2000 hours or 12 months shelf life.
 View more versions of this product
Catalog No. 14-500-728
Add to cart
Description
Description
  • Long life
  • Drop-in OEM replacement
  • Traceable by serial number
Order Info
Cat. No. 14-500-728 is comparable to Varian part number 391615691.
Compliance
WEEE and ROHS compliant
Specifications
Specifications
Varian UV50/100/200 9050 Prostar 31 UV and Visible HPLC Detector
Deuterium
Replacement Detector Lamp