Learn More
BrandTech™ Replacement Discharge Tube Screw Caps for BRAND Dispensette S Bottletop Dispensers
Screw caps and hinged strap for use with Dispensette S models made 2016 and later.
Manufacturer:  BrandTech™ 706025
 View more versions of this product
Catalog No. 14-380-886
Add to cart
Description
Description
  • 706018 is for Dispensette S (red trim) dispensers with a nominal (max) capacity of 10mL or smaller
  • 706019 is for Dispensette S (red trim) dispensers with a nominal (max) capacity of 25mL or larger
  • 706025 is for Dispensette S Organic (yellow trim) dispensers with a nominal (max) capacity of 5mL or 10mL
  • 706027 is for Dispensette S Organic (yellow trim) dispensers with a nominal (max) capacity of 25mL or larger
  • 706029 is for Dispensette S Trace Analysis (white/clear trim) dispensers, or Dispensette S or S Organic models with a nominal capacity of 10mL or smaller and enhanced chemical compatibility is required
  • 706031 is for Dispensette S and S Organimc models with a nominal capacity of 25mL or larger, and enhanced chemical compatibility is required.
Specifications
Specifications
Discharge tube screw cap and fastener for BRAND Dispensette S Organic models with a nominal capacity of 5 or 10mL
Yes
BRAND Dispensette S Organic
1
Yellow