Replacement Drums for Eppendorf™ Cell Culture Roller Drum

Manufacturer: Eppendorf North America ATC-TT16

 View all options for this product

Catalog No. 14-277-4


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Type Roller Drum
    Diameter (English) 14 in.
    Diameter (Metric) 35.6cm
    For Use With (Equipment) New Brunswick cell culture roller drum
    Holds 164 Tubes of 16mm O.D.
    Material Aluminum

    For 16 and 25mm tubes. Lightweight, 14 in. diameter (35.6cm) aluminum drums, hold 164 tubes up to 16mm diameter, or 80 tubes up to 25mm diameter. Drums are easily removable, can be used as carrying trays.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit