Learn More
Idex Replacement Ferrules for SealTight™ Fittings
Slotted end on ferrule creates second compression zone for a tight seal
Manufacturer:  Idex F192
 View more versions of this product
Catalog No. 05-701-375
Add to cart
Specifications
Specifications
10-32 or M6 coned, for 1/16 in. OD tubing
PEEK
Black
1/Ea
Ferrule