Learn More
Fisherbrand™ Replacement Filters for PCR Workstation
Prefilters for use with PCR workstations.
 View more versions of this product
Catalog No. 103-603-10
Add to cart
Description
Description
  • Replacement disposable prefilters made of corrugated paper trap large particles to extend HEPA filter life
  • Recommend replacing the prefilter every three months
  • For use with 2 ft. and 3 ft. PCR Enclosures
Specifications
Specifications
Filter
3 ft. PCR workstation
2.54 cm
28.5 in.
16.5 in.
16.5 x 28.5 in. (41.91 x 72.4 cm)
Disposable Prefilter
1 in.
72.4 cm
Corrugated Paper
41.91 cm