Learn More
Thermo Scientific™ Replacement Module for Orion™ Nitrate Sure-Flow Combination Electrode PROMO
Measure nitrate in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Nitrate Electrodes.
Manufacturer:  Thermo Scientific™ 970701PM
Catalog No. 13-301-11PM
Add to cart
Description
Description
  • Calibration standards catalog number: 920706 - 0.1m NaNO3; 920707 - 1000 ppm N; 930707 - 100 ppm N
  • Required ISA catalog number: 930711 or 930710 / nitrate ISS
Specifications
Specifications
Combination Ion Selective Electrode
dri ft.-free potentials
ISEmeters
Replacementmodule for Nitrate Sure-Flow Combination Electrode 9707 BNWP; 1/Pk.
Free-flowing Sure-FlowTM junction assures stable