Learn More
Thermo Scientific™ Replacement Needles for LC Syringes Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ Replacement Needles for LC Syringes are available for syringes with removable needles.
Manufacturer:  Thermo Scientific™ 365RNL15
 View more versions of this product
Catalog No. 36-5RN-L15
  Check Availability
Add to cart
Specifications
Description
Thermo Scientific™ Replacement Needles for LC Syringes are available for syringes with removable needles.
Description & Specifications
Specifications
365RNL15
2 in.
Syringe Needle
10µL
LC Autosampler Syringe
50.8mm
22 ga.