Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trajan™ Replacement Needle for Syringe with Valve
Quality needles to fit an SGE syringe prefitted with removable valves
Supplier: Trajan™ 038815
Description
Replacement Needles for use with syringes prefitted with removable valves
- Needle Length: 50mm
- Needle Gauge: 23
- OD: 0.63mm
- ID: 0.2mm
Choose Cone or Side Hole/Dome tip style
Specifications
23 G | |
0.63 mm | |
Side Hole or Dome |
0.20 mm | |
Replacement Valve Needle |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title