Learn More
MilliporeSigma™ Supelco™ Replacement Stainless Steel Filter - 7 µm
For use with Gas Line Filters
$29.85 - $31.95
Specifications
Type Replacement Filter
Material Stainless Steel
For Use With (Application) Replacing Worn or Damaged Filters
View More Specs
Products 2
Catalog Number Mfr. No. For Use With Price Quantity & Availability  
Catalog Number Mfr. No. For Use With Price Quantity & Availability  
11-101-6127
View Documents
MilliporeSigma™ Supelco™
25810U
1/8 in. Gas Line Filter (Cat. No. 20620)
Each for $31.95
Add to cart
11-100-5153
View Documents
MilliporeSigma™ Supelco™
25809
1/4 in. Gas Line Filter (Cat. No. 20621)
Each for $29.85
Add to cart
Description
Description
  • Use to replace worn or damaged filters in a gas line filter
  • Not compatible with T-type gas line filters
Specifications
Specifications
Replacement Filter
Stainless Steel
Replacing Worn or Damaged Filters