Learn More
Trajan™ SilTite™ FingerTite Ferrule
For gas chromatography systems
$91.80
Specifications
Type GC Ferrules
Quantity 10/Pk.
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (Metric) Inside Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Inside Price Quantity & Availability  
SG073632
View Documents
Trajan™
073632
0.7 mm
Pack of 10 for $91.80
Add to cart
SG073631
View Documents
Trajan™
073631
0.5 mm
Pack of 10 for $91.80
Add to cart
SG073630
View Documents
Trajan™
073630
0.4 mm
Pack of 10 for $91.80
Add to cart
Description
Description
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Specifications
GC Ferrules
10/Pk.