Thermo Scientific™ Accessories and Replacement Parts for MaxiMix™ III Vortex Mixer
Optimize flexibility of the Thermo Scientific™ MaxiMix™ III Vortex Mixer with this selection of accessories and replacement parts.
Manufacturer: Thermo Scientific™ PT500X9A
 View more versions of this product
Catalog No. 12-815-1D
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    For Use With (Equipment) MaxiMix™ III Vortex Mixer
    Type Universal Holder
    Description Universal holder for a variety of flasks, beakers, bottles, test tub racks. Two soft rubber bars hold vessels in place.
    For Use With For the MaxiMix III Vortex Mixer for optimum flexibility
    Utilize these Thermo Scientific™ Accessories and Replacement Parts for MaxiMix™ III Vortex Mixer to optimize flexibility of the machine.
  • Description & Specifications
Related Products
Product Content Feedback