Learn More
Research Products International CorpSupplier Diversity Partner Sterile Replacement Filter for RPI Pipette Filler, 0.2um DFS Item
Manufacturer:  Research Products International CorpSupplier Diversity Partner 248648
The RPI Pipette Filler fits all glass or plastic pipettes from 1-100ml. Features eight speeds for aspirating and dispensing liquids with high accuracy. Lightweight ergonomic design allows one hand operation and reduces pipetting fatigue. Individual controls are provided for aspirate and dispense operations. Fills a 50ml pipette in less than five seconds and dispenses liquid by gravity flow or motorized operation. Built in Lithium-ion battery provides a long run time between charges. LCD panel displays speed selection and battery charge. Supplied with two hydrophobic 0.45 um filters and a three year warranty.
Catalog No. 50-476-558