Learn More
MP Biomedicals™ Replica Plating Apparatus (1 Unit)
Replica Plating Apparatus (1 Unit)
Manufacturer:  MP Biomedicals™ 115000001
Catalog No. MP115000001
Add to cart
Description
Description
  • Safely and easily spread yeast and bacterial cells
  • Spread cells in just seconds per plate
  • Eliminate hazards of fl ame and ethanol
  • No need for fragile glass "e;hockey sticks”
  • Consistent, even colony distribution
  • Sterile and easy to use
  • Made from hardwood
  • the Replica Plating Apparatus can be used with velvet, gauze, or even paper towels to transfer colonies from plate to plate
  • or plate to fi lter while retaining colony orientation.
  • The Replica Plating Apparatus and Velvet Squares can be ordered to fit either 100 mm or 150
  • mm plates.
Specifications
Specifications
Wooden Sphere with Block of woodwith Hole for Sphere
Bacteria Cell Culture; Cell Preservation; Cell Counting, Viability, and Cryopreservation; Insect Cell Culture; Mammalian Cell Culture; Plant Cellgrowth; Primary Cell Culture; Stem Cell Culture; Yeast Cell Culture
100 mm