Learn More
REPS2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 206991AP
Catalog No. 50-173-3042
Add to cart
Description
Description
The antibody is specific to REPS2.
The product of this gene is part of a protein complex that regulates the endocytosis of growth factor receptors. The encoded protein directly interacts with a GTPase activating protein that functions downstream of the small G protein Ral. Its expression can negatively affect receptor internalization and inhibit growth factor signaling. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
REPS2
0.56 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q80XA6, Q8NFH8
Partner of RalBP1, POB1, RalBP1 interacting protein 2, REPS2
Rabbit
IgG
150 μL
-20°C
Primary
194590, 9185
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
REPS2
Liquid
REPS2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse